Skip to Content
Merck
All Photos(2)

Key Documents

417696

Sigma-Aldrich

Zinc bis[bis(trimethylsilyl)amide]

greener alternative

97%

Synonym(s):

Bis[1,1,1-trimethyl-N-(trimethylsilyl)silanaminato]zinc, Bis[bis(trimethylsilyl)amido]zinc, Zinc bis(hexamethyldisilazide, 1,1,1,3,3,3-Hexamethyldisilazane zinc salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[[(CH3)3Si]2N]2Zn
CAS Number:
Molecular Weight:
386.16
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

liquid

reaction suitability

core: zinc
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

12.5 °C (lit.)

density

0.957 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

C[Si](C)(C)N([Zn]N([Si](C)(C)C)[Si](C)(C)C)[Si](C)(C)C

InChI

1S/2C6H18NSi2.Zn/c2*1-8(2,3)7-9(4,5)6;/h2*1-6H3;/q2*-1;+2

InChI key

ZCCUYQBZUVUONI-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application


  • Cooperative effects of Schiff base binuclear zinc complexes on the synthesis of aliphatic and semi-aromatic polyesters.: This study explores the cooperative effects of binuclear zinc complexes in the synthesis of various polyesters, highlighting the potential of zinc bis[bis(trimethylsilyl)amide] in enhancing polymerization efficiency and selectivity (Santulli et al., 2023).

  • Synthesis, solution dynamics and chemical vapour deposition of heteroleptic zinc complexes via ethyl and amide zinc thioureides.: The authors discuss the synthesis and dynamics of zinc complexes for chemical vapor deposition, emphasizing the role of zinc bis[bis(trimethylsilyl)amide] as a precursor in developing high-purity metalorganic films (Bhide et al., 2021).

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service