Skip to Content
Merck
All Photos(1)

Key Documents

332895

Sigma-Aldrich

Trichloromethanesulfenyl chloride

≥96%

Synonym(s):

Perchloromethyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CCl3SCl
CAS Number:
Molecular Weight:
185.89
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.41 (vs air)

vapor pressure

2.02 psi ( 20 °C)

Assay

≥96%

form

liquid

refractive index

n20/D 1.537 (lit.)

bp

146-148 °C (lit.)

density

1.7 g/mL at 25 °C (lit.)

SMILES string

ClSC(Cl)(Cl)Cl

InChI

1S/CCl4S/c2-1(3,4)6-5

InChI key

RYFZYYUIAZYQLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trichloromethanesulfenyl chloride was used in the preparation of:
  • trichloromethanesulfenyl acetate and trichloromethanesulfenyl trifluoroacetate via heterogeneous reaction with silver acetate and silver trifluoroacetate
  • trichloromethyl disulfides
  • alkyl orthocarbonates
  • trithiocarbonates

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Some reactions of trichloromethanesulfenyl chloride with alcohols and thiols.
Islam NB, et al.
Journal of Chemical and Engineering Data, 30(4), 509-512 (1985)
Lin Du et al.
The journal of physical chemistry. A, 111(23), 4944-4949 (2007-05-22)
Two novel species, trichloromethanesulfenyl acetate, CCl(3)SOC(O)CH(3), and trichloromethanesulfenyl trifluoroacetate, CCl(3)SOC(O)CF(3), have been generated in situ by the heterogeneous reactions between trichloromethanesulfenyl chloride, CCl(3)SCl, and corresponding silver salts, silver acetate (AgOC(O)CH(3)) and silver trifluoroacetate (AgOC(O)CF(3)), respectively. Photoelectron spectroscopy and quantum chemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service