Skip to Content
Merck
All Photos(1)

Key Documents

268976

Sigma-Aldrich

3-Methyl-1,3-pentadiene, mixture of cis and trans

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(CH3)CH=CH2
CAS Number:
Molecular Weight:
82.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.451 (lit.)

bp

75-77 °C (lit.)

density

0.73 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C\C=C(/C)C=C

InChI

1S/C6H10/c1-4-6(3)5-2/h4-5H,1H2,2-3H3/b6-5+

InChI key

BOGRNZQRTNVZCZ-AATRIKPKSA-N

Application

3-Methyl-1,3-pentadiene was used in the synthesis of two major nitronic ester cycloadducts by the tin(IV)-catalyzed reaction with β-nitrostyrene in toluene[1].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter A Wade et al.
Beilstein journal of organic chemistry, 9, 2137-2146 (2013-11-10)
The tin(IV)-catalyzed reaction of β-nitrostyrene with (E)-3-methyl-1,3-pentadiene in toluene afforded two major nitronic ester cycloadducts in 27% and 29% yield that arise from the reaction at the less substituted diene double bond. Also present were four cycloadducts from the reaction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service