Skip to Content
Merck
All Photos(2)

Documents

228605

Sigma-Aldrich

3-Chloro-4-methoxyaniline

97%

Synonym(s):

3-Chloro-p-anisidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(OCH3)NH2
CAS Number:
Molecular Weight:
157.60
Beilstein:
879162
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

50-55 °C (lit.)

SMILES string

COc1ccc(N)cc1Cl

InChI

1S/C7H8ClNO/c1-10-7-3-2-5(9)4-6(7)8/h2-4H,9H2,1H3

InChI key

XQVCBOLNTSUFGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Chloro-4-methoxyaniline is aniline metabolite of chlorpropham and has been quantitated by by micellar electrokinetic capillary chromatography with laser-induced fluorescence detection.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva Orejuela et al.
Electrophoresis, 26(15), 2991-2998 (2005-07-05)
A rapid, reliable method has been developed for the multi-residue analysis of aniline metabolites of chlorpropham in potato samples. The method involves the precolumn derivatization of aniline metabolites with 5-(4,6-dichloro-s-triazin-2-ylamino) fluorescein (DTAF) and their subsequent separation and determination by micellar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service