225088
5-Amino-2-methylphenol
97%
Synonym(s):
3-Hydroxy-4-methylaniline, 4-Amino-2-hydroxytoluene, 5-Amino-o-cresol
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
H2NC6H3(CH3)OH
CAS Number:
Molecular Weight:
123.15
Beilstein:
2802317
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
160-162 °C (lit.)
SMILES string
Cc1ccc(N)cc1O
InChI
1S/C7H9NO/c1-5-2-3-6(8)4-7(5)9/h2-4,9H,8H2,1H3
InChI key
DBFYESDCPWWCHN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The spectrophotometric course of the reaction of 5-amino-2-methylphenol with p-benzoquinone di-imine was studied.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Benzoquinone imines. Part IX. Mechanism and kinetics of the reaction of p-benzoquinone di-imines with m-aminophenols.
J. Chem. Soc. Perkin Trans. II, 5, 539-548 (1972)
Allergic contact dermatitis caused by cysteamine hydrochloride in permanent wave agent-A new allergen for hairdressers in Japan.
Kazue Nishioka et al.
Contact dermatitis, 80(3), 174-175 (2018-12-12)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service