Skip to Content
Merck
All Photos(2)

Key Documents

182680

Sigma-Aldrich

Poly(vinyl formal)

powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

refractive index

n20/D 1.502

transition temp

Tg 108 °C

density

1.23 g/mL at 25 °C (lit.)

SMILES string

O(COC=C)C=C

InChI

1S/C5H8O2/c1-3-6-5-7-4-2/h3-4H,1-2,5H2

InChI key

DFFZYNIDEOOVAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service