Skip to Content
Merck
All Photos(3)

Documents

155861

Sigma-Aldrich

4-Fluoro-3-nitroaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(NO2)NH2
CAS Number:
Molecular Weight:
156.11
Beilstein:
2210199
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

94-96 °C (lit.)

SMILES string

Nc1ccc(F)c(c1)[N+]([O-])=O

InChI

1S/C6H5FN2O2/c7-5-2-1-4(8)3-6(5)9(10)11/h1-3H,8H2

InChI key

LLIOADBCFIXIEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Fluoro-3-nitroaniline was used in the preparation of commercial hair dyes.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Barbee et al.
Toxicology and applied pharmacology, 72(3), 400-405 (1984-03-15)
4-Fluoro-3-nitroaniline (4-F-3-NA), an intermediate used in the production of commercial hair dyes, was administered daily for 90 days via po intubation to Charles River CD rats at doses of 40, 120, or 360 mg/kg to determine its subchronic toxicology. All
S Chen et al.
Journal of neurochemistry, 45(3), 940-945 (1985-09-01)
We had previously shown that 4-fluoro-3-nitrophenyl azide (FNPA) is a competitive inhibitor of both types of monoamine oxidase (MAO) in the dark, but it is a preferential photoaffinity label for only the type B MAO (MAO-B). Recently we synthesized a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service