Skip to Content
Merck
All Photos(1)

Key Documents

155586

Sigma-Aldrich

3-Fluoro-4-methoxybenzaldehyde

99%

Synonym(s):

3-Fluoro-p-anisaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(OCH3)CHO
CAS Number:
Molecular Weight:
154.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

34-35 °C (lit.)

functional group

aldehyde

SMILES string

COc1ccc(C=O)cc1F

InChI

1S/C8H7FO2/c1-11-8-3-2-6(5-10)4-7(8)9/h2-5H,1H3

InChI key

SOQCZBSZZLWDGU-UHFFFAOYSA-N

General description

The nuclear magnetic resonance spectral parameters for 3-Fluoro-4-methoxybenzaldehyde has been studied.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The range of in anisole derivatives. An indicator of torsion and compression of the methoxy group.
Canadian Journal of Chemistry, 63(3), 782-786 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service