154385
2-Thiophenecarbonitrile
99%
Synonym(s):
2-Cyanothiophene, Thiophene-2-carbonitrile
About This Item
Assay
99%
form
liquid
refractive index
n20/D 1.563 (lit.)
bp
192 °C (lit.)
density
1.172 g/mL at 25 °C (lit.)
SMILES string
N#Cc1cccs1
InChI
1S/C5H3NS/c6-4-5-2-1-3-7-5/h1-3H
InChI key
CUPOOAWTRIURFT-UHFFFAOYSA-N
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
127.4 °F - closed cup
Flash Point(C)
53 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service