All Photos(3)
About This Item
Linear Formula:
CH3(CH2)7Cl
CAS Number:
Molecular Weight:
148.67
Beilstein:
1697464
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
1 mmHg ( 22.2 °C)
Quality Level
Assay
99%
form
liquid
refractive index
n20/D 1.430 (lit.)
bp
183 °C (lit.)
mp
−61 °C (lit.)
density
0.875 g/mL at 25 °C (lit.)
functional group
alkyl halide
chloro
SMILES string
CCCCCCCCCl
InChI
1S/C8H17Cl/c1-2-3-4-5-6-7-8-9/h2-8H2,1H3
InChI key
CNDHHGUSRIZDSL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
1-Chlorooctane has been used to determine the excess molar enthalpies of binary mixtures composed by 2-alkanone and 1-chloroalkane. It has been used in hydrophobic modification of ω-carrageenans polymers.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
141.8 °F - closed cup
Flash Point(C)
61 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Excess enthalpies of some 2-alkanone+ 1-chloroalkane binary mixtures at 25 and 35? C.
Pico JM, et al.
Journal of Solution Chemistry, 20(1), 115-124 (1991)
Hydrophobically modified calcium-induced ?-carrageenan gels.
Jacquier JC, et al.
Prog. Coll. Polym. Sci., 118, 127-131 (2001)
Biosynthesis and characterization of a new bacterial copolyester of 3-hydroxyalkanoates and 3-hydroxy-omega-chloroalkanoates.
Doi Y and Abe C.
Macromolecules, 23(15), 3705-3707 (1990)
Susann Wegner et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(26), 6330-6340 (2017-02-15)
The microwave-induced decomposition of bis{N,N'-diisopropylacetamidinate}nickel(II) [Ni{MeC(NiPr)
A R King et al.
British journal of pharmacology, 157(6), 974-983 (2009-06-03)
Monoacylglycerol lipase (MGL) is a presynaptic serine hydrolase that inactivates the endocannabinoid neurotransmitter, 2-arachidonoyl-sn-glycerol. Recent studies suggest that cysteine residues proximal to the enzyme active site are important for MGL function. In the present study, we characterize the role of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service