Merck
  • Cross-coupling of cyclopropyl- and cyclobutyltrifluoroborates with aryl and heteroaryl chlorides.
Cross-coupling of cyclopropyl- and cyclobutyltrifluoroborates with aryl and heteroaryl chlorides.
The Journal of organic chemistry (2008-09-02)
Gary A Molander, Paul E Gormisky
ABSTRACT
Suitable conditions were found for the Suzuki-Miyaura cross-coupling reaction of potassium cyclopropyl- and cyclobutyltrifluoroborates with aryl chlorides. Both of these secondary alkyl trifluoroborates coupled in moderate to excellent yield with electron-rich, electron-poor, and hindered aryl chlorides to give a variety of substituted aryl cyclopropanes and cyclobutanes.
Sigma-Aldrich
Cyclobutylboronic acid, ≥95.0%