- Recombinant toxins for cancer treatment.
Recombinant toxins for cancer treatment.
Science (New York, N.Y.) (1991-11-22)
I Pastan, D FitzGerald
PMID1683495
ABSTRACT
Recombinant toxins target cell surface receptors and antigens on tumor cells. They kill by mechanisms different from conventional chemotherapy, so that cross resistance to conventional chemotherapeutic agents should not be a problem. Furthermore, they are not mutagens and should not induce secondary malignancies or accelerate progression of benign malignancies. They can be mass-produced cheaply in bacteria as homogeneous proteins. Either growth factor-toxin fusions or antibody-toxin fusions can be chosen, depending on the cellular target.
Sigma-Aldrich
Anti-Pseudomonas aeruginosa Exotoxin A antibody, Mouse monoclonal, clone EXO-68, purified from hybridoma cell culture