
(S)-4-(4-Aminobenzyl)-2(1H)-oxazolidinone

Synonym(s):
(4S)-4-[(4-Aminophenyl)methyl]-2-oxazolidinone
Empirical Formula (Hill Notation):
C10H12N2O2
CAS Number:
Molecular Weight:
192.21
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Package Size
Special Grade
Showing 1-2 of 2