1076206
USP
Powdered Black Cohosh Extract
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Cimicifuga racemosa extract
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
black cohosh
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- Black Cohosh
- Black Cohosh Fluidextract
- Black Cohosh Tablets
- Powdered Black Cohosh
- Powdered Black Cohosh Extract
Analysis Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service