Skip to Content
Merck
All Photos(1)

Key Documents

PHR9030

Supelco

N-Acetylbenzocaine

reference material

Synonym(s):

Ethyl 4-acetamidobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO3
CAS Number:
Molecular Weight:
207.23
MDL number:
UNSPSC Code:
41116107

grade

reference material

API family

benzocaine

Assay

≥97.0% (HPLC)

form

solid

shelf life

limited shelf life, expiry date on the label

application(s)

pharmaceutical small molecule

storage temp.

2-8°C

InChI

1S/C11H13NO3/c1-3-15-11(14)9-4-6-10(7-5-9)12-8(2)13/h4-7H,3H2,1-2H3,(H,12,13)

InChI key

NHLOBHNRBWKNIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service