Skip to Content
Merck
All Photos(1)

Key Documents

807646

TSKgel® QAE-2SW (5 µm) HPLC Columns

L x I.D. 2.5 cm × 6 mm, ammonium ion, quaternary phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
L × I.D.:
2.5 cm × 6 mm
particle size:
5 μm
matrix active group:
ammonium ion, quaternary phase
matrix:
spherical silica

Product Name

TSKgel® QAE-2SW Guardgel Kit, phase ammonium ion, quaternary, L × I.D. 2.5 cm × 6 mm, 5 μm particle size

material

stainless steel

Agency

suitable for USP L14

product line

TSKgel®

feature

counter ion H2PO4-

manufacturer/tradename

Tosoh 07646

parameter

0-100% organic solvent
10-45 °C temperature

technique(s)

HPLC: suitable

L × I.D.

2.5 cm × 6 mm

matrix

spherical silica

matrix active group

ammonium ion, quaternary phase

particle size

5 μm

operating pH

2.0-7.5

separation technique

anion exchange

General description

TSKgel Guardgel kits contain:
  • 5 mL Guardgel packing material
  • Column holder, consisting of external reducing union, cap, frit, 2 ferrules
  • Column, 25 mm x 9.5 mm OD x 6 mm ID, 2 μm frit at 10 mm depth, 1/16" exit
  • 2 Replacement frits
  • Capillary tubing, 5 cm x 1/16" OD x 0.01" ID
  • Nuts and ferrules, 10-32 threads for 1/16" tubing, 2 each
  • all hardware is 316 stainless steel

Physical form

Shipped in methanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service