Key Documents
58458
Supelco
Waters® Fittings
size 1/16 in., pkg of 5 ea
About This Item
UNSPSC Code:
41115700
Pricing and availability is not currently available.
description
nuts and ferrules (5 ea)
packaging
pkg of 5 ea
size
1/16 in.
General description
The Waters® Fittings are used for connecting most HPLC columns to a 1/16 in. tubing system.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Waters is a registered trademark of Waters
Choose from one of the most recent versions:
Certificates of Analysis (COA)