Skip to Content
Merck
All Photos(1)

Key Documents

500429

Supelco

JHAP-43 Mix

1 ppm in nitrogen, pkg of 110 L (pi-marked cylinder), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12000000

grade

analytical standard

Agency

EPA TO-14

packaging

pkg of 110 L (pi-marked cylinder)

manufacturer/tradename

Scott Gas T9Z90000-110

concentration

1 ppm in nitrogen

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

General description

JHAP-43 Mix is used as an air monitoring standard, which employs TO-14 SUMA canister sampling for GC/MS analysis of volatile organics in ambient air.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Acrylonitrile
    Allyl chloride
    Benzene
    Bromomethane
    1,3-Butadiene
    Carbon tetrachloride
    Chlorobenzene
    Chloroethane
    Chloroform
    Chloromethane
    1,2-Dibromoethane
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    Dichlorodifluoromethane
    1,1-Dichloroethane
    1,2-Dichloroethane
    1,1-Dichloroethylene
    cis-1,2-Dichloroethylene
    Dichloromethane
    1,2-Dichloropropane
    cis-1,3-Dichloropropene
    trans-1,3-Dichloropropene
    1,2-Dichlorotetrafluoroethane
    Ethylbenzene
    4-Ethyltoluene
    Hexachloro-1,3-butadiene
    Styrene
    1,1,2,2-Tetrachloroethane
    Tetrachloroethylene
    Toluene
    1,2,4-Trichlorobenzene
    1,1,1-Trichloroethane
    1,1,2-Trichloroethane
    Trichloroethylene
    Trichlorofluoromethane
    1,1,2-Trichloro-1,2,2-trifluoroethane
    1,2,4-Trimethylbenzene
    1,3,5-Trimethylbenzene
    Vinyl chloride
    o-Xylene
    m-Xylene
    p-Xylene
    See All (43)

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service