Key Documents
33195
Supelco
Crimpers
Hand crimper, for use with 11mm crimp seals, pkg of 1 ea
About This Item
UNSPSC Code:
41122001
packaging
pkg of 1 ea
compatibility
for use with 11mm crimp seals
General description
The Supelco hand held crimper provides a consistent and dependable seal that allows secure vial closure each and evey time. The high quality construction is engineered for durability and long life to provide a smooth and simple operation.
Application
Hand crimper may be used to seal a glass vial containing MeOH, for high-performance liquid chromatography-diode array detection (HPLC-DAD) analysis, during polyurethanes (PUs) analysis and clean-up, in an experimental study done in order to develop, optimise and apply PU as new polymeric phases for stir bar sorptive extraction.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
N R Neng et al.
Journal of chromatography. A, 1171(1-2), 8-14 (2007-10-12)
In this contribution, polyurethane foams are proposed as new polymeric phases for stir bar sorptive extraction (SBSE). Assays performed for polyurethane synthesis demonstrated that four series of formulations (P(1), P(2), P(3) and P(4)) present remarkable stability and excellent mechanical resistance