Skip to Content
Merck
All Photos(1)

Key Documents

21131-U

Supelco

Purge/Trap K Vocarb® 3000

for use with OI Analytical 4460

Synonym(s):

Vocarb® 3000 Purge Trap K, O.I. Analytical

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NB.21
Pricing and availability is not currently available.

application(s)

environmental

compatibility

for use with OI Analytical 4460

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vocarb is a registered trademark of Siemens Industry, Inc.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service