Z744132
TiterSeal Sealing Film
polyester, W × L 3.125 in. × 5.750 in.
Synonym(s):
adhesive sealing film, adhesive sealing tape, microplate sealing film, microplate sealing tape, sealing film, sealing tape
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.35
material
polyester
sterility
non-sterile
packaging
pkg of 100 ea
manufacturer/tradename
Excel Scientific TTS-100
parameter
-40-+120 °C temp. range
technique(s)
PCR: suitable
W × L
3.125 in. × 5.750 in.
General description
Minimizes evaporation, prevents well-to-well contamination and spillage
- Eliminates the “edge effect” which can occur withplate lids (e.g. greater evaporation in the outsideplate wells)
- Provides a secure seal and is easy to remove
- Rigid polyester film features good clarity and easyhandling
- Can be used as a temporary economical seal whileawaiting further processing
- Features two end-tabs with perforations
Application
Ideal for General laboratory use, short-term storage, incubation and ELISA
Features and Benefits
Minimizes evaporation, prevents well-to-well contamination and spillage
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service