Key Documents
Z708593
Hexagonal weighing boats
size L
Synonym(s):
weighboat, weiging dish
About This Item
UNSPSC Code:
41121812
NACRES:
NB.71
Pricing and availability is not currently available.
material
hexagonal
polystyrene
size
L
feature
antistatic
packaging
pack of 500 ea
manufacturer/tradename
Heathrow Scientific HS14251C
bottom I.D.
3 in. (76 mm)
top I.D.
4 in. (102 mm)
General description
  • Hexagonal weighing boats are ideal for handling samples for weighing.
  • Flexible boats can be easily bent to form pour spouts for spill-free transfer and prevent sample loss.
  • Flat bottoms and sloping slides provide an accurate transfer.
  • These antistatic, polystyrene boats are non-reactive to most substances.
  • Chemically resistant to most chemicals including weak acids, aqueous solutions, alcohols, and bases.
  • Compatible with microwave moisture analyzers.
  • Resist temperatures to 158 °F (70 °C).
Features and Benefits
  • Available in four different sizes and 1 color
  • Pack of 500
  • FDA/ food grade compliant products
  • Disposable
Choose from one of the most recent versions:
Certificates of Analysis (COA)