Z380350
Slant rack for culture tubes
Holds 40 x culture tubes
Synonym(s):
rack test tube, racks tube, test tube racks
All Photos(1)
About This Item
UNSPSC Code:
41122804
NACRES:
NB.22
Recommended Products
feature
Holds 40 x culture tubes
packaging
pack of 1 ea
case of 4 ea
manufacturer/tradename
Nalgene® 5935-0020
General description
Unique end plate design allows 5° or 20° slant when rack is set down on side. Autoclave media upright, then allow to cool on a side. Media will harden with perfect, consistent slant. Also useful for incubating liquids on a slant for better aeration.
Rack holds 40 tubes (4 × 10 array); keyed with numbers and letters. Stackable; will not float in water bath. Autoclaving will reduce strength over time.
Rack holds 40 tubes (4 × 10 array); keyed with numbers and letters. Stackable; will not float in water bath. Autoclaving will reduce strength over time.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)