Skip to Content
Merck
All Photos(1)

Key Documents

UC211

Sigma-Aldrich

7-Hydroxywarfarin

Synonym(s):

3-(α-Acetonylbenzyl)-4,7-dihydrocoumarin, 4-7-Dihydroxy-3-(3-oxo-1-phenylbutyl)-2H-1-benzopyran-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16O5
CAS Number:
Molecular Weight:
324.33
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

mp

220 °C

solubility

DMSO: soluble

storage temp.

2-8°C

SMILES string

CC(CC(C1=C(O)C(C=CC(O)=C2)=C2OC1=O)C3=CC=CC=C3)=O

InChI

1S/C19H16O5/c1-11(20)9-15(12-5-3-2-4-6-12)17-18(22)14-8-7-13(21)10-16(14)24-19(17)23/h2-8,10,15,21-22H,9H2,1H3

InChI key

SKFYEJMLNMTTJA-UHFFFAOYSA-N

Application

7-Hydroxywarfarin can be used for the metabolic assays of (S)-warfarin.

Biochem/physiol Actions

CYP2C9 metabolite of (S)-warfarin.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

7-Hydroxywarfarin is soluble in DMSO.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tae Inoue et al.
Drug metabolism and disposition: the biological fate of chemicals, 36(12), 2429-2433 (2008-09-12)
Chimeric mice having humanized livers were constructed by transplantation of human hepatocytes. In this study, we investigated whether these mice have a capacity for drug metabolism similar to that of humans by examining hydroxylation of S-warfarin, which is predominantly metabolized
Masahiro Utoh et al.
Biochemical pharmacology, 95(2), 110-114 (2015-03-25)
Cynomolgus monkeys are widely used as non-human primate species in preclinical studies, due to their close evolutionary relationship to humans. Monkey cytochrome P450 2C19 (formerly known as P450 2C75), highly homologous to human P450 2C19, has been identified to be
D Lang et al.
Journal of chromatography. B, Biomedical applications, 672(2), 305-309 (1995-10-20)
The formation of 7-hydroxywarfarin in incubations of (S)-warfarin with human liver microsomes reflects their cytochrome P-4502C9 activity. This paper describes a rapid high-performance liquid chromatographic method for the determination of 7-hydroxywarfarin with high sensitivity, selectivity, and a simple sample clean-up
Xuemin Jiang et al.
British journal of clinical pharmacology, 57(5), 592-599 (2004-04-20)
M: The aim of this study was to investigate the effect of St John's wort and ginseng on the pharmacokinetics and pharmacodynamics of warfarin. This was an open-label, three-way crossover randomized study in 12 healthy male subjects, who received a
D J Black et al.
Drug metabolism and disposition: the biological fate of chemicals, 24(4), 422-428 (1996-04-01)
Consistent with expectations based on human in vitro microsomal experiments, administration of fluconazole (400 mg/day) for 6 days to six human volunteers significantly reduced the cytochrome P450 (P450)-dependent metabolic clearance of the warfarin enantiomers. In particular, P4502C9 catalyzed 6- and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service