Skip to Content
Merck
All Photos(1)

Key Documents

SRP6499

Sigma-Aldrich

HAVCR1/KIM1/TIM1 human

recombinant, expressed in HEK 293 cells, ≥90% (SDS-PAGE)

Synonym(s):

HAVCR, HAVCR-1, Hepatitis A virus cellular receptor 1, TIMD-1, TIMD1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥90% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 31.3 kDa
observed mol wt 80-100 kDa by SDS-PAGE (reducing) (DTT-reduced.)

packaging

pkg of 10 and 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HAVCR1(26762)

General description

Hepatitis A virus cellular receptor 1 also known as HAVCR1, HAVCR, KIM1, TIM, TIM1, TIMD1, is widely expressed with highest levels in kidney and testis. The protein encoded by HAVCR1 gene is a membrane receptor for both human hepatitis A virus (HHAV) and TIMD4. The encoded protein may be involved in the moderation of asthma and allergic diseases. The reference genome represents an allele that retains a MTTVP amino acid segment that confers protection against atopy in HHAV seropositive individuals. Three transcript variants encoding the same protein have been found for this gene. HAVCR1 may play a role in T-helper cell development, the regulation of asthma and allergic diseases and in kidney injury and repair. In case of human hepatitis A virus (HHAV) infection, functions as a cell-surface receptor for the virus.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service