SRP5314
CK1γ1, active, GST tagged human
PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
CK1G1, CKI-gamma 1, CSNK1G1
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
recombinant
expressed in baculovirus infected Sf9 cells
product line
PRECISIO®
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
66-98 nmol/min·mg
mol wt
~82 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... CSNK1G1(53944)
General description
CK1γ1 or casein kinase 1, gamma 1 is belonging to the most abundant serine/threonine kinase in eukaryotic cell extracts which is mainly involved in growth and morphogenesis of eukaryotic cells. Casein kinase 1, gamma 1 possesses a C-terminal most terminal sequence motif (MTM), which it shares with CSNK1G2 and CSNK1G3 which is associated with heterologous carboxy-terminal sequences. Casein kinase 1-gamma couples Wnt receptor activation to the cytoplasmic signal transduction apparatus.
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.
Preparation Note
After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service