Skip to Content
Merck
All Photos(1)

Key Documents

SML3624

Sigma-Aldrich

Cefditoren pivoxil

≥98% (HPLC)

Synonym(s):

(6R,7R)-7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-3-[(1Z)-2-(4-methyl-5-thiazolyl)ethenyl]-8-oxo-5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid (2,2-dimethyl-1-oxopropoxy)methyl ester, CCRIS 7768, ME 1207

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H28N6O7S3
CAS Number:
Molecular Weight:
620.72
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

[s]1c(nc(c1)\C(=N\OC)\C(=O)N[C@H]2[C@H]3SCC(=C(N3C2=O)C(=O)OCOC(=O)C(C)(C)C)\C=C/c4[s]cnc4C)N

InChI

1S/C25H28N6O7S3/c1-12-15(41-10-27-12)7-6-13-8-39-21-17(29-19(32)16(30-36-5)14-9-40-24(26)28-14)20(33)31(21)18(13)22(34)37-11-38-23(35)25(2,3)4/h6-7,9-10,17,21H,8,11H2,1-5H3,(H2,26,28)(H,29,32)/b7-6-,30-16-/t17-,21-/m1/s1

InChI key

AFZFFLVORLEPPO-UVYJNCLZSA-N

Biochem/physiol Actions

Cefditoren pivoxil is a prodrug of an orally available, broad spectrum third-generation cephalosporin antibiotic Cefditoren. It is active against both gram-negative and gram-positive bacteria. Cefditoren pivoxil is used for treatment of chronic bronchitis, community-acquired pneumonia, pharyngitis/tonsillitis, and uncomplicated skin and skin-structure infections.
Prodrug of an orally available, broad spectrum third-generation cephalosporin antibiotic Cefditoren.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taiga Miyazaki et al.
Journal of infection and chemotherapy : official journal of the Japan Society of Chemotherapy, 25(9), 702-707 (2019-04-16)
Oral antibiotic therapy for patients with acute exacerbations of chronic obstructive pulmonary disease (COPD) usually involves an aminopenicillin with clavulanic acid, a macrolide, or a quinolone. To date, however, the clinical efficacy and safety of the oral cephalosporin cefditoren pivoxil
Effectiveness of antibacterial agents against cell-invading bacteria such as Streptococcus pyogenes and Haemophilus influenzae
BMC Microbiology, 21(1), 148-148 (2021)
A Tamura et al.
Antimicrobial agents and chemotherapy, 32(9), 1421-1426 (1988-09-01)
ME1207 (pivaloyloxymethyl ester of ME1206) is a new oral cephalosporin. ME1206 is (6R,7R)-7-[(Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)- acetamido]-3-[(Z)-2-(4-methylthiazol-5-yl)-ethyl]-cephem-4-carboxy lic acid. The susceptibilities of about 1,600 clinical isolates to ME1206 were determined by the agar dilution method. ME1206 showed a broad spectrum of activity against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service