Skip to Content
Merck
All Photos(1)

Key Documents

SML3371

Sigma-Aldrich

MS47134

≥98% (HPLC)

Synonym(s):

((1r,3R,5S,7R)-3,5-dimethyladamantane-1-carbonyl)-D-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29NO3
Molecular Weight:
355.47
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N[C@H](CC1=CC=CC=C1)C(O)=O)[C@]2(C[C@]3(C)C4)C[C@@H]4C[C@](C3)(C)C2

InChI

1S/C22H29NO3/c1-20-9-16-10-21(2,12-20)14-22(11-16,13-20)19(26)23-17(18(24)25)8-15-6-4-3-5-7-15/h3-7,16-17H,8-14H2,1-2H3,(H,23,26)(H,24,25)/t16-,17-,20+,21-,22-/m1/s1

InChI key

QSBRCNGPJBWGLQ-LQDXOGNASA-N

Biochem/physiol Actions

MS47134 is a potent and selective agonist probe of the GPCR orphan receptor MRGPRX4.
Potent and selective agonist probe of the GPCR orphan receptor MRGPRX4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Can Cao et al.
Nature, 600(7887), 170-175 (2021-11-19)
The MRGPRX family of receptors (MRGPRX1-4) is a family of mas-related G-protein-coupled receptors that have evolved relatively recently1. Of these, MRGPRX2 and MRGPRX4 are key physiological and pathological mediators of itch and related mast cell-mediated hypersensitivity reactions2-5. MRGPRX2 couples to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service