Skip to Content
Merck
All Photos(1)

Key Documents

SML3341

Sigma-Aldrich

ML-SA5

≥98% (HPLC)

Synonym(s):

N1-(3-Chloro-2-(piperidin-1-yl)phenyl)-N4,N4-dimethylbenzene-1,4-disulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24ClN3O4S2
CAS Number:
Molecular Weight:
457.99
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

Biochem/physiol Actions

ML-SA5 is a potent TRPML cation channel agonist that activates whole-endolysosomal TRPML1 (ML1) currents in DMD myoblasts at a higher potency than ML-SA1 (EC50 = 285 nM vs 10 μM). ML-SA5 ameliorates muscular dystrophy in mdx, but not ML1-knockout, mice in vivo (2 or 5 mg/kg/day i.p.) by facilitating sarcolemma repair, thereby reducing skeletal and cardiac muscle damage with no apparent toxicity to the animals.
Potent TRPML1 cation channel agonist that ameliorates muscular dystrophy in mdx, but not ML1-KO, mice and with a higher potency than ML-SA1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service