Skip to Content
Merck
All Photos(1)

Key Documents

SML3338

Sigma-Aldrich

NF-56-EJ40

≥98% (HPLC)

Synonym(s):

2-(2-(4′-((4-Methylpiperazin-1-yl)methyl)biphenyl-3-ylcarboxamido)phenyl)acetic acid, 2-(2-(4′-((4-methylpiperazin-1-yl)methyl)-[1,1′-biphenyl]-3-carboxamido)phenyl)acetic acid, 2-[[[4′-[(4-Methyl-1-piperazinyl)methyl][1,1′-biphenyl]-3-yl]carbonyl]amino]-benzeneacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29N3O3
CAS Number:
Molecular Weight:
443.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

NF-56-EJ40 is a high-affinity, potent and human species-selective succinate receptor SUCNR1 (GPR91) antagonist (human GPR91 Ki/KD = 17.4/33 nM; GTPγS assay IC50 = 25 nM/human, >100 μM/rat). Extracellular succinate is reported to hyperpolarize M2 macrophages through GPR91-mediated Gq signaling, blockable by NF-56-EJ40 (1 μM) or Gq inhibitor YM-254890 (1 μM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service