Skip to Content
Merck
All Photos(1)

Key Documents

SML2832

Sigma-Aldrich

5-Amino-1-methylquinolinium iodide

≥98% (HPLC)

Synonym(s):

5-Amino-1-MQ iodide, 5-Amino-1-methylquinolinium, iodide (1:1), 5-Amino-1MQ iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N2·I
CAS Number:
Molecular Weight:
286.11
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

faint red to dark brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

C[N+]1=CC=CC2=C(C=CC=C12)N.[I-]

InChI

1S/C10H10N2.HI/c1-12-7-3-4-8-9(11)5-2-6-10(8)12;/h2-7,11H,1H3;1H

InChI key

JPEZFBFIRRAFNR-UHFFFAOYSA-N

Biochem/physiol Actions

5-Amino-1-methylquinolinium (5-amino-1MQ) is a substrate site-targeting, selective nicotinamide N-methyltransferase (NNMT) inhibitor (IC50 = 1.2 μM; 50 μM SAM, 100 μM NCA) that reduces 3T3-L1 lipogenesis (EC50 = 30μM) and adipocytes 1-methylnicotinamide level (EC50 = 2.3 μM) without affecting related methyltransferases & enzymes in the NAD+ salvage pathway. 5-amino-1MQ shows in vivo therapeutic efficacy in murine models of DIO (20 mg/kg/d sc.), muscle injury (5 or 10 mg/kg bid. sc.), and intraperitoneal HeyA8 ovarian cancer metastasis (20 mg/kg/d ip.) with no adverse effects to the animals (up to 60 mg/kg/d). Note:1.89 mg iodide salt contains 1 mg eq of 5-amino-1MQ.
Substrate site-targeting, selective nicotinamide N-methyltransferase (NNMT) inhibitor with in vitro and in vivo efficay.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harshini Neelakantan et al.
Journal of medicinal chemistry, 60(12), 5015-5028 (2017-05-27)
Nicotinamide N-methyltransferase (NNMT) is a fundamental cytosolic biotransforming enzyme that catalyzes the N-methylation of endogenous and exogenous xenobiotics. We have identified small molecule inhibitors of NNMT with >1000-fold range of activity and developed comprehensive structure-activity relationships (SARs) for NNMT inhibitors.
Mark A Eckert et al.
Nature, 569(7758), 723-728 (2019-05-03)
High-grade serous carcinoma has a poor prognosis, owing primarily to its early dissemination throughout the abdominal cavity. Genomic and proteomic approaches have provided snapshots of the proteogenomics of ovarian cancer1,2, but a systematic examination of both the tumour and stromal
Harshini Neelakantan et al.
Biochemical pharmacology, 147, 141-152 (2017-11-21)
There is a critical need for new mechanism-of-action drugs that reduce the burden of obesity and associated chronic metabolic comorbidities. A potentially novel target to treat obesity and type 2 diabetes is nicotinamide-N-methyltransferase (NNMT), a cytosolic enzyme with newly identified
Harshini Neelakantan et al.
Biochemical pharmacology, 163, 481-492 (2019-02-13)
Aging is accompanied by progressive declines in skeletal muscle mass and strength and impaired regenerative capacity, predisposing older adults to debilitating age-related muscle deteriorations and severe morbidity. Muscle stem cells (muSCs) that proliferate, differentiate to fusion-competent myoblasts, and facilitate muscle

Questions

1–2 of 2 Questions  
  1. Can there be a chloride salt version that is white?

    1 answer
    1. Unfortunately, the iodide form of this compound is the only product option available at this time.

      Helpful?

  2. Does 5 amino 1MQ have iodine in it? 

    1 answer
    1. Yes, iodine is present as this product is the iodide salt of 5-Amino-1MQ . This is not a covalent bond and the Iodine will ionize in solution. Please see the link below to review a sample Certificate of Analysis:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/SML2832-5MG0000102634.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service