Skip to Content
Merck
All Photos(1)

Key Documents

SML1547

Sigma-Aldrich

RN-9893

≥98% (HPLC)

Synonym(s):

N-(4-(4-Isopropylpiperazin-1-ylsulfonyl)phenyl)-2-nitro-4-(trifluoromethyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23F3N4O5S
CAS Number:
Molecular Weight:
500.49
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=C(S(=O)(N2CCN(C(C)C)CC2)=O)C=C1)C3=CC=C(C(F)(F)F)C=C3[N+]([O-])=O

InChI

1S/C21H23F3N4O5S/c1-14(2)26-9-11-27(12-10-26)34(32,33)17-6-4-16(5-7-17)25-20(29)18-8-3-15(21(22,23)24)13-19(18)28(30)31/h3-8,13-14H,9-12H2,1-2H3,(H,25,29)

InChI key

KORKKQLDGMHNKO-UHFFFAOYSA-N

General description

RN-9893 is a small, orally bioavailable antagonist of TRPV4 (transient receptor potential subfamily V4. It is a tri-substituted benzamide compound.

Biochem/physiol Actions

RN-9893 is a potent and selective antagonist of the Transient Receptor Potential ion channel TRPV4. RN-9893 has IC50 values of 420 nM, 660 nM, and 320 nM for human, rat and mouse TRPV4 receptors, respectively. RN-9893 shows excellent selectivity over related TRP receptors with no inhibition of TRPV1 at a concentration of 10 μM, an IC50 >30 μM against TRPV3, an IC50 of approximately 30 μM against TRPM8, and also good selectivity using 54 binding assays against common biological targets.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of orally-bioavailable antagonists of the TRPV4 ion-channel.
Wei ZL et al
BioTechnology: An Indian Journal, 25(18), 4011-4015 (2015)
TRPV4 agonists and antagonists.
Vincent F and Duncton MA
Current Topics in Medicinal Chemistry, 11(17), 2216-2226 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service