Skip to Content
Merck
All Photos(1)

Key Documents

SML0842

Sigma-Aldrich

CRANAD 2

≥98% (HPLC)

Synonym(s):

(T-4)-[(1E,6E)-1,7-bis[4-(Dimethylamino)phenyl]-1,6-heptadiene-3,5-dionato-?O3,?O5]difluoroboron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25BF2N2O2
CAS Number:
Molecular Weight:
410.26
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

, dark blue to black

solubility

DMSO: 0.2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[B-]1([O+]=C(C=C(O1)\C=C\c3ccc(cc3)N(C)C)\C=C\c2ccc(cc2)N(C)C)(F)F

InChI

1S/C23H25BF2N2O2/c1-27(2)20-11-5-18(6-12-20)9-15-22-17-23(30-24(25,26)29-22)16-10-19-7-13-21(14-8-19)28(3)4/h5-17H,1-4H3/b15-9+,16-10+

InChI key

PVXQJYXODFZTBR-KAVGSWPWSA-N

Biochem/physiol Actions

CRANAD 2 is a near-infrared (NIR) Aβ plaque-specific fluorescent probe.
CRANAD 2 is a near-infrared (NIR) Aβ plaque-specific fluorescent probe. CRANAD 2 penetrates the blood brain barrier and has a high affinity for Aβ aggregates with a Kd of 38 nM. Upon interacting with Aβ aggregates, CRANAD 2 exhibits a 70-fold increase in fluorescence intensity accompanied by a 90 nm blue shift from 805 to 715 nm and a large increase in quantum yield. CRANAD 2 has been used to detect Aβ deposits noninvasively in vivo in transgenic Tg2576 mice.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service