Skip to Content
Merck
All Photos(1)

Documents

SML0444

Sigma-Aldrich

Pluripotin

≥98% (HPLC)

Synonym(s):

N-[3-[7-(2,5-Dimethyl-2H-pyrazol-3-ylamino)-1-methyl-2-oxo-1,4-dihydro-2H-pyrimido[4,5-d]pyrimidin-3-yl]-4-methylphenyl]-3-trifluoromethyl-benzamide, SC 1, SC-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H25F3N8O2
CAS Number:
Molecular Weight:
550.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL (clear solution)

storage temp.

−20°C

SMILES string

CN1C(=O)N(Cc2cnc(Nc3cc(C)nn3C)nc12)c4cc(NC(=O)c5cccc(c5)C(F)(F)F)ccc4C

InChI

1S/C27H25F3N8O2/c1-15-8-9-20(32-24(39)17-6-5-7-19(11-17)27(28,29)30)12-21(15)38-14-18-13-31-25(34-23(18)36(3)26(38)40)33-22-10-16(2)35-37(22)4/h5-13H,14H2,1-4H3,(H,32,39)(H,31,33,34)

InChI key

NBZFRTJWEIHFPF-UHFFFAOYSA-N

Biochem/physiol Actions

Pluripotin is an activator of murine embryonic stem (ES) cell self-renewal. It appears that pluripotin mediates the activity by dual RasGAP and ERK1 inhibition.

Features and Benefits

This compound is featured on the MAPKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ye F Tian et al.
Biomaterials, 73, 110-119 (2015-09-26)
Non-Hodgkin lymphomas are a heterogeneous group of lymphoproliferative disorders of B and T cell origin that are treated with chemotherapy drugs with variable success rate that has virtually not changed over decades. Although new classes of chemotherapy-free epigenetic and metabolic
Alejandro Gil-Gálvez et al.
Proceedings of the National Academy of Sciences of the United States of America, 119(11), e2114802119-e2114802119 (2022-03-10)
SignificanceIn this manuscript, we address an essential question in developmental and evolutionary biology: How have changes in gene regulatory networks contributed to the invertebrate-to-vertebrate transition? To address this issue, we perturbed four signaling pathways critical for body plan formation in
Franziska Hentzschel et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 22(12), 2130-2141 (2014-09-06)
Malaria, caused by protozoan Plasmodium parasites, remains a prevalent infectious human disease due to the lack of an efficient and safe vaccine. This is directly related to the persisting gaps in our understanding of the parasite's interactions with the infected

Articles

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service