Skip to Content
Merck
All Photos(3)

Key Documents

SMB00703

Sigma-Aldrich

Tetraacetylphytosphingosine

≥98% (HPLC)

Synonym(s):

N-[(1S,2S,3R)-2,3-bis(acetyloxy)-1-[(acetyloxy)methyl]heptadecyl]-acetamide, TAPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H47NO7
CAS Number:
Molecular Weight:
485.65
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

mp

46 °C

solubility

ethanol: ≤25 mg/mL

lipid type

sphingolipids

storage temp.

−20°C

SMILES string

N([C@H]([C@H](OC(=O)C)[C@H](OC(=O)C)CCCCCCCCCCCCCC)COC(=O)C)C(=O)C

InChI

1S/C26H47NO7/c1-6-7-8-9-10-11-12-13-14-15-16-17-18-25(33-22(4)30)26(34-23(5)31)24(27-20(2)28)19-32-21(3)29/h24-26H,6-19H2,1-5H3,(H,27,28)/t24-,25+,26-/m0/s1

InChI key

SGTYQWGEVAMVKB-NXCFDTQHSA-N

Biochem/physiol Actions

Tetraacetylphytosphingosine (TAPS) is a sphingolipid metabolite produced by Wickerhamomyces ciferrii (also known as Hansenula ciferrii).[1] It exerts an inhibitory action on angiogenesis through inhibition of the mitogen-activated protein kinase (MAPK) pathway and intracellular calcium increase. Therefore, TAPS was suggested as a potential treatment for angiogenesis related disorders such as arthritis, cancer and psoriasis.[2]

TAPS was found to induce apoptosis in HaCaT human keratinocyte cells. In addition, it was demonstrated that TAPS synergistically increases UVB-induced apoptosis via caspase activation by regulating the level of pro-apoptotic Bax and anti-apoptotic Bcl-2 in HaCaT cells.[3]

Other Notes

Due to the low transition temperature of tetraacetylphytosphingosine, the product may be a semi-solid at room temperature.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoo Bin Kwon et al.
Experimental dermatology, 16(4), 311-317 (2007-03-16)
In a search for the wound healing accelerators, we found that tetraacetyl-phytosphingosine (TAPS), a sphingolipid metabolite produced by phytosphingosine acetylation, has significant inhibitory potential on healing of rabbit ear wound. As angiogenesis is fundamental to proper wound healing, we examined
H J Kim et al.
Apoptosis : an international journal on programmed cell death, 9(4), 449-456 (2004-06-12)
Inappropriate apoptosis results in the epidermal hyperplasia as in psoriasis and UVB irradiation has been successfully used to treat this kind of skin disorders. Previously, we reported that the novel phytosphingosine derivative, tetraacetyl phytosphingosine (TAPS) induced apoptosis in HaCaT cells.
Studies on the production of sphingolipid bases by the yeast, Hansenula ciferri.
M L Greene et al.
Biochimica et biophysica acta, 98(3), 582-588 (1965-06-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service