SIC003
MISSION® siRNA Fluorescent Universal Negative Control #1, Cyanine 3
Synonym(s):
MISSION® siRNA Fluorescent Universal Negative Control, Cyanine 3
About This Item
Recommended Products
product line
MISSION®
color
Green-Yellow (Cyanine 3)
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Application
Features and Benefits
- Cyanine 3 labeled siRNA Universal Negative Control allows easy visualization of the siRNA in your cells by using fluorescence microscopy.
- Rapidly assess transfection efficiency.
- Ready to use.
- Available in three sizes for your convenience.
- Compatible with Human, Mouse, and Rat cells.
Legal Information
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
MISSION siRNA Negative Controls ensure mRNA knockdown efficiency baseline; tested in human, rat, and mouse cells, designed without gene homology.
MISSION siRNA Negative Controls ensure mRNA knockdown efficiency baseline; tested in human, rat, and mouse cells, designed without gene homology.
MISSION siRNA Negative Controls ensure mRNA knockdown efficiency baseline; tested in human, rat, and mouse cells, designed without gene homology.
MISSION siRNA Negative Controls ensure mRNA knockdown efficiency baseline; tested in human, rat, and mouse cells, designed without gene homology.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service