Skip to Content
Merck
All Photos(1)

Documents

SBPV04

Sigma-Aldrich

BCRP Human Vesicular Transport Kit

membrane based kit for Human BCRP Vesicular Transport Assays

Synonym(s):

ABCG2, Breast Cancer Resistance Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

recombinant

expressed in human cells

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ABCG2(9429)

General description

SOLVO Biotechnology offers ready-to-use assay kits for efflux and uptake transporters. SOLVO′s PREDIVEZ Vesicular Transport Assay Kits are suitable for efflux transporter inhibition studies. The kits are designed to test the inhibitory effect of one or more test compounds on the vesicular transport of a probe substrate. In most cases a fluorescent probe substrate is included. Kits contain all reagents and solutions necessary to conduct the drug-transporter interaction assay of need.

Application

The PREDIVEZTM Vesicular Transport Kit is a simple and powerful tool to investigate drug transporter interactions. It provides information on any interaction between the ABC transporter and the drug candidate that would affect the transport of the fluorescent or radiolabeled or cold.

PREDIVEZTM vesicular transport kits are available in three sizes 3, 6 or 9 compound sizes. The kits contain the sufficient amount of reagents and membrane preparations needed for the testing of 3 compounds per 96-well plate. Each kit is accompanied by a CD that includes the assay protocol, an Excel spreadsheet with a data processing template, SDS and the inhibition curves with reference compounds.

Physical form

Supplied as frozen membrane vesicles, containing 5 mg/ml membrane protein, labeled with volume, catalog number (transporter), date of production, protocols and necessary reagents sufficent for the analysis of 3, 6, or 9 test compounds

Legal Information

Distributed for SOLVO Biotechnology, Inc.

related product

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Lact. - Met. Corr. 1 - Repr. 1A - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Albert Mennone et al.
Drug metabolism and disposition: the biological fate of chemicals, 38(10), 1673-1678 (2010-07-06)
Breast cancer resistance protein (Bcrp) is a member of the ATP-binding cassette membrane transporter family, which is expressed apically in liver, kidney, and intestine epithelium. Recent reports suggest that in addition to xenobiotics, porphyrins, and food toxins, Bcrp can also
M Maliepaard et al.
Cancer research, 61(8), 3458-3464 (2001-04-20)
High expression of the Breast Cancer Resistance Protein (BCRP) gene has been shown to be involved in resistance to chemotherapeutic drugs. Knowledge of the localization of BCRP protein in normal tissues may help unravel the normal function of this protein.
Márton Jani et al.
Biological & pharmaceutical bulletin, 32(3), 497-499 (2009-03-03)
The pharmacokinetics of sulfasalazine, an anti-inflammatory drug is influenced by ATP-binding cassette G2 (ABCG2) (breast cancer resistance protein (BCRP), mitoxantrone resistance protein (MXR)) both in vitro and clinically. Due to its low passive permeability, the intracellular concentration of sulfasalazine is
Hiroshi Kodaira et al.
The Journal of pharmacology and experimental therapeutics, 333(3), 788-796 (2010-03-23)
A synergistic effect of P-glycoprotein (P-gp)/Abcb1a and breast cancer resistance protein (Bcrp)/Abcg2 was reported to limit the brain penetration of their common substrates. This study investigated this based on pharmacokinetics using Mdr1a/1b(-/-), Bcrp(-/-), and Mdr1a/1b(-/-)/Bcrp(-/-) mice. Comparison of the brain-
Kumie Kage et al.
International journal of cancer, 97(5), 626-630 (2002-01-25)
Breast cancer resistance protein (BCRP) is a half-molecule ABC transporter highly expressed in mitoxantrone-resistant cells. In our study we established PA317 transfectants expressing Myc-tagged BCRP (MycBCRP) or HA-tagged BCRP (HABCRP). The exogenous BCRP protein migrated as a 70-kDa protein in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service