Skip to Content
Merck
All Photos(1)

Key Documents

RAB1378

Sigma-Aldrich

Human CMG-2 ELISA

for serum, plasma and cell culture supernatants

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ANTXR2(118429)

General description

Anthrax toxin receptor (ANTXR2), also known as capillary morphogenesis protein gene-2 (CMG2), is encoded by the gene mapped to human chromosome 24q21. CMG2 is a ubiquitously expressed type I transmembrane protein. It contains von Willebrand A (vWA) domain that is involved in binding lamin and collagen IV.
This ELISA antibody pair detects Human CMG-2. Other species not yet determined.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Biochem/physiol Actions

Anthrax toxin receptor (ANTXR2)/ capillary morphogenesis protein gene-2 (CMG2) has been implicated in basement-membrane matrix assembly and endothelial cell morphogenesis. CMG2 acts as a chief receptor for anthrax in vivo. It stimulates the anthrax toxin activity by equipping toxin with a high-affinity binding anchor on the cell membrane and a path of entry into the host cell. Mutation in the gene is associated with the development of rare, autosomal recessive disorders called, Juvenile hyaline fibromatosis (JHF) and infantile systemic hyalinosis (ISH).

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Systemic hyalinosis with heterozygous CMG2 mutations: a case report and review of literature.
Rahvar M, et al.
The American Journal of Dermatopathology, 38(5), e60-e63 (2016)
Identification of 2 Novel ANTXR2 Mutations in Patients With Hyaline Fibromatosis Syndrome and Proposal of a Modified Grading System
Denadai R, et al.
American Journal of Medical Genetics. Part A, 158(4), 732-742 (2012)
Roles of Anthrax Toxin Receptor 2 in Anthrax Toxin Membrane Insertion and Pore Formation
Sun J, et al.
Toxins, 8(2), 34-34 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service