Skip to Content
Merck
All Photos(1)

Key Documents

RAB0680

Sigma-Aldrich

Human IGF2R / Cation-independent mannose-6-phosphate Receptor ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.77

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.27 ng/mL
standard curve range: 0.274-200 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IGF2R(3482)

General description

The antibody pair provided in this kit recognizes Human Insulin-Like Growth Gactor type II Receptor.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Vicikova et al.
Neoplasma, 65(5), 762-768 (2018-06-27)
The mannose 6-phosphate/insulin-like growth factor 2 receptor (CD222, M6P/IGF2R) is a multifunctional transmembrane type I receptor, mostly localized intracellularly, less on the surface of all types of mammalian cells. It is known both to transport lysosomal enzymes through their mannose
Brian Dwyer et al.
Protein expression and purification, 170, 105589-105589 (2020-02-07)
The cation-independent mannose-6-phosphate receptor (CI-M6PR, aka insulin-like growth factor II receptor or IGFIIR) is a membrane protein that plays a central role in the trafficking of lysosomal acid hydrolases into lysosomes via mannose-6-phosphate (M6P) binding domains. In order to maintain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service