Skip to Content
Merck
All Photos(1)

Key Documents

PZ0296

Sigma-Aldrich

PF-06459988

≥98% (HPLC)

Synonym(s):

1-[(3R,4R)-3-[[[5-Chloro-2-[(1-methyl-1H-pyrazol-4-yl)amino]-7H-pyrrolo[2,3-d]pyrimidin-4-yl]oxy]methyl]-4-methoxy-1-pyrrolidinyl]-2-propen-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22ClN7O3
CAS Number:
Molecular Weight:
431.88
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear

storage temp.

room temp

SMILES string

ClC1=CNC2=C1C(OC[C@@H]3[C@@H](OC)CN(C(C=C([H])[H])=O)C3)=NC(NC4=CN(C)N=C4)=N2

InChI

1S/C19H22ClN7O3/c1-4-15(28)27-7-11(14(9-27)29-3)10-30-18-16-13(20)6-21-17(16)24-19(25-18)23-12-5-22-26(2)8-12/h4-6,8,11,14H,1,7,9-10H2,2-3H3,(H2,21,23,24,25)/t11-,14+/m1/s1

InChI key

ODMXWZROLKITMS-RISCZKNCSA-N

Biochem/physiol Actions

PF-06459988 is an orally available irreversible inhibitor of epidermal growth factor receptor (EGFR) mutant (EGFRm) forms including the secondary acquired resistance mutation T790M. PF-06459988 has minimal activity against wild-type EGFR (WT EGFR).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hengmiao Cheng et al.
Journal of medicinal chemistry, 59(5), 2005-2024 (2016-01-13)
First generation EGFR TKIs (gefitinib, erlotinib) provide significant clinical benefit for NSCLC cancer patients with oncogenic EGFR mutations. Ultimately, these patients' disease progresses, often driven by a second-site mutation in the EGFR kinase domain (T790M). Another liability of the first

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service