Skip to Content
Merck
All Photos(1)

Key Documents

PZ0267

Sigma-Aldrich

WAY-362450

≥98% (HPLC)

Synonym(s):

3-(3,4-Difluoro-benzoyl)-1,1-dimethylene-1,2,3,6-tetrahydro-azepino [4,5-b]indole-5-carboxylic acid isopropyl ester, 3-(3,4-Difluorobenzoyl)-1,2,3,6-tetrahydro-1,1-dimethyl-azepino[4,5-b]indole-5-carboxylic acid 1-methylethyl ester, FXR 450, Turofexorate isopropyl, WAY 362450, XL335

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24F2N2O3
CAS Number:
Molecular Weight:
438.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

FC1=C(F)C=C(C(N2CC(C)(C)C(C(C=CC=C3)=C3N4)=C4C(C(OC(C)C)=O)=C2)=O)C=C1

InChI

1S/C25H24F2N2O3/c1-14(2)32-24(31)17-12-29(23(30)15-9-10-18(26)19(27)11-15)13-25(3,4)21-16-7-5-6-8-20(16)28-22(17)21/h5-12,14,28H,13H2,1-4H3

InChI key

INASOKQDNHHMRE-UHFFFAOYSA-N

Gene Information

human ... NR1H4(9971)

Biochem/physiol Actions

WAY-362450 is a potent and selective Farnesoid X receptor (FXR) agonist that lowers total plasma cholesterol (all lipoprotein species). WAY-362450 reduced the levels of high-density lipoprotein cholesterol (HDLc) in Cynomolgus monkeys, mice and hamsters. Apparently HDL lowering is achieved through increased transhepatic cholesterol efflux.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service