Skip to Content
Merck
All Photos(1)

Documents

PZ0175

Sigma-Aldrich

PD 149163 tetrahydrochloride hydrate

≥90% (HPLC)

Synonym(s):

N2-[(2S)-2,6-Diaminohexyl]-L-lysyl-L-prolyl-L-tryptophyl-3-methyl-L-valyl-L-Leucine ethyl ester tetrahydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H71N9O6·4HCl · xH2O
CAS Number:
Molecular Weight:
943.91 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

powder (Hygroscopic)

storage condition

desiccated

color

white to tan

solubility

H2O: ≥12 mg/mL

storage temp.

2-8°C

SMILES string

O.Cl.Cl.Cl.Cl.CCOC(=O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@H](Cc1c[nH]c2ccccc12)NC(=O)[C@@H]3CCCN3C(=O)[C@H](CCCCN)NC[C@@H](N)CCCCN)C(C)(C)C

InChI

1S/C42H71N9O6.4ClH.H2O/c1-7-57-41(56)34(23-27(2)3)49-39(54)36(42(4,5)6)50-37(52)33(24-28-25-46-31-17-9-8-16-30(28)31)48-38(53)35-19-14-22-51(35)40(55)32(18-11-13-21-44)47-26-29(45)15-10-12-20-43;;;;;/h8-9,16-17,25,27,29,32-36,46-47H,7,10-15,18-24,26,43-45H2,1-6H3,(H,48,53)(H,49,54)(H,50,52);4*1H;1H2/t29-,32-,33-,34-,35-,36+;;;;;/m0...../s1

InChI key

PIFVEGOSSWCHFK-AEPODNBTSA-N

Biochem/physiol Actions

PD 149163 is a Neurotensin NTR1 receptor agonist; a Neurotensin (8-13) analog. PD149163 is a selective, brain penetrating NT1 receptor agonist with pro-cognitive, antipsychotic and anxiolytic effects.

Features and Benefits

This compound is featured on the Neurotensin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam J Prus et al.
Progress in neuro-psychopharmacology & biological psychiatry, 49, 78-84 (2013-11-28)
Neurotensin is an endogenous neuropeptide that has significant interactions with monoamine neurotransmitter systems. To date, neurotensin NTS1 receptor agonists, such as PD149163, have been primarily evaluated for the treatment for schizophrenia, drug addiction, and pain. Recently, PD149163 was found to
Elizabeth N Holly et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 21(7), 526-531 (2011-02-01)
Agonists for neurotensin (NT)-1 receptors have produced antipsychotic-like effects in many animals, including reversal of prepulse inhibition deficits and psychostimulant-induced increases in spontaneous activity. The present study sought to provide a basic assessment of the putative antipsychotic effects of PD149163
Todd M Hillhouse et al.
Experimental and clinical psychopharmacology, 22(6), 548-556 (2014-10-07)
Major depressive disorder is the most common mood disorder in the United States and European Union; however, the limitations of clinically available antidepressant drugs have led researchers to pursue novel pharmacological treatments. Clinical studies have reported that monotherapy with the

Articles

Neurotensin is a 13 amino acid regulatory peptide found mainly in gut and brain. Intestinal neurotensin is produced by a discrete population of endocrine cells (N cells) scattered throughout the jejuno-ileal mucosa.

Neurotensin is a 13 amino acid regulatory peptide found mainly in gut and brain. Intestinal neurotensin is produced by a discrete population of endocrine cells (N cells) scattered throughout the jejuno-ileal mucosa.

Neurotensin is a 13 amino acid regulatory peptide found mainly in gut and brain. Intestinal neurotensin is produced by a discrete population of endocrine cells (N cells) scattered throughout the jejuno-ileal mucosa.

Neurotensin is a 13 amino acid regulatory peptide found mainly in gut and brain. Intestinal neurotensin is produced by a discrete population of endocrine cells (N cells) scattered throughout the jejuno-ileal mucosa.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service