Skip to Content
Merck
All Photos(2)

Key Documents

PRS3959

Sigma-Aldrich

Anti-Slug (ab2) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Neural crest transcription factor slug, Anti-Snail homolog 2 (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 29 kDa

species reactivity

human, mouse

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNAI2(6591)

Immunogen

a 13 amino acid peptide from near the center of human Slug.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3959.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenny Chitcholtan et al.
International journal of molecular sciences, 25(8) (2024-04-27)
Ovarian cancer poses a significant threat to patients in its advanced stages, often with limited treatment options available. In such cases, palliative management becomes the primary approach to maintaining a reasonable quality of life. Therefore, the administration of any medication
Qiwei Yang et al.
PloS one, 18(6), e0286454-e0286454 (2023-06-23)
The black garlic is produced from the raw garlic by Milliard reaction at high temperature (~60-90°C) and humidity (~70-90%). In this process, the pungent odor and gastrointestinal irritation effects of the raw garlic are reduced. At the same time, unstable
Neha Ahuja et al.
NAR cancer, 2(3), zcaa021-zcaa021 (2020-10-23)
Hypoxic microenvironment heralds epithelial-mesenchymal transition (EMT), invasion and metastasis in solid tumors. Deregulation of alternative splicing (AS) of several cancer-associated genes has been instrumental in hypoxia-induced EMT. Our study in breast cancer unveils a previously unreported mechanism underlying hypoxia-mediated AS
Ziqian Yan et al.
Cell death & disease, 12(12), 1120-1120 (2021-12-03)
Studies have shown that cancer-associated fibroblasts (CAFs) play an irreplaceable role in the occurrence and development of tumors. Therefore, exploring the action and mechanism of CAFs on tumor cells is particularly important. In this study, we compared the effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service