Skip to Content
Merck
All Photos(1)

Key Documents

P4651

Sigma-Aldrich

Promethazine hydrochloride

Synonym(s):

10-[2-(Dimethylamino)propyl]phenothiazine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2S · HCl
CAS Number:
Molecular Weight:
320.88
Beilstein:
4166397
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

form

powder

originator

Wyeth

storage temp.

2-8°C

SMILES string

Cl[H].CC(CN1c2ccccc2Sc3ccccc13)N(C)C

InChI

1S/C17H20N2S.ClH/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H

InChI key

XXPDBLUZJRXNNZ-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

H1 histamine receptor antagonist; CNS depressant; anticholinergic.

Features and Benefits

This compound was developed by Wyeth. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Other Notes

Solubility: Freely soluble in water, in hot dehydrated alcohol and in Chloroform. Practically insoluble in ether, in acetone and in ethyl acetate.

Physical form

Appearance: White or faint yellow, odorless, Crystalline powder, slowly oxidizes and acquires a blue color, on prolonged exposure to air.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A B Wright et al.
Ultrasound in medicine & biology, 25(2), 241-247 (1999-05-13)
This sonographic study was aimed at examining the effect of sedation with promethazine (1.5 mg x kg(-1)), on gastroduodenal function in neonatal piglets. On 3 consecutive days, observations of gastroduodenal motility during the first 3 postprandial h were made in
Z Li et al.
Neuroscience, 70(1), 145-158 (1996-01-01)
Ionic mechanisms responsible for histamine-induced prolonged depolarization in supraoptic nucleus neurons were investigated using whole-cell patch recordings in horizontally prepared brain slices from adult male rats. Bath application of histamine (1-10 microM) in control medium induced membrane depolarization in nine
Frank Fasbender et al.
Archives of toxicology, 94(2), 439-448 (2020-02-16)
Drug-induced liver injury (DILI) represents one of the major causes why drugs have to be withdrawn from the market. In this study, we describe a new interaction between drug-exposed hepatocytes and natural killer (NK) cells. In a previous genome-wide expression
William C Kwan et al.
The Journal of comparative neurology, 527(3), 558-576 (2018-01-03)
The primate visual brain possesses a myriad of pathways, whereby visual information originating at the retina is transmitted to multiple subcortical areas in parallel, before being relayed onto the visual cortex. The dominant retinogeniculostriate pathway has been an area of
Louis C Leung et al.
Nature, 571(7764), 198-204 (2019-07-12)
Slow-wave sleep and rapid eye movement (or paradoxical) sleep have been found in mammals, birds and lizards, but it is unclear whether these neuronal signatures are found in non-amniotic vertebrates. Here we develop non-invasive fluorescence-based polysomnography for zebrafish, and show-using unbiased, brain-wide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service