Skip to Content
Merck
All Photos(1)

Documents

N60

Sigma-Aldrich

Triton N-60

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161900
NACRES:
NA.25

description

non-ionic

Quality Level

form

liquid

General description

Triton N-60 is a planar aromatic species with an approximate length of 10 Å.

Application

Triton N-60 has been used in a study to assess the use of surfactant solutions for dust suppression in coal mines. It has also been used in a study to describe a spectrophotometric assay for determination of critical micelle concentration of nonionic detergent solutions.

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Critical micelle concentration determination of nonionic detergents with Coomassie Brilliant Blue G-250
Ken S. Rosenthal and Frank Koussaie
Analytical Chemistry, 55, 1115-1115 (1983)
Molecular Rectangles Based on Rhenium(I) Coordination Chemistry
Kurt D. Benkstein et al.
Journal of the American Chemical Society, 120, 12982-12982 (1998)
Coal wetting ability of surfactant solutions and the effect of multivalent anion additions
Howard W. Kilau and John E. Pahlman.
Colloids Surf., 26, 217-217 (1987)
Yanfeng Liu et al.
Toxicology in vitro : an international journal published in association with BIBRA, 65, 104778-104778 (2020-01-22)
The need of in vitro alternative methods has been increasing in toxicology research as well as in cosmetic industry in China recently. Following the establishment of China EpiSkin™ skin corrosion and irritation testing methods, both as stand-alone in vitro tests

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service