MTCB13008BK
MTC™ Bio MultiBox™ Western Blot Box Pull-off Lid
L × W × H 8.6 cm × 8.6 cm × 2.8 cm
Synonym(s):
gel processing box lid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
Recommended Products
material
polystyrene
packaging
pack of 10 ea
manufacturer/tradename
MTC™ Bio B1300-8BK
L × W × H
8.6 cm × 8.6 cm × 2.8 cm
General description
Blot Box, Removable Lid, Opaque Black, for Novex Minigel (8.6 x 8.6 x 2.8cm), 10/pk
Application
Novex Mini Gels
Features and Benefits
- A perfect box for every blot
- Reduce antibody usage
- Hinged lid minimizes evaporation
- For halves, wholes, strips or minis
Legal Information
MTC is a trademark of MTC Bio
MultiBox is a trademark of MTC Bio
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service