Skip to Content
Merck
All Photos(1)

Documents

M6890

Sigma-Aldrich

Muscle acetone powder from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

biological source

rabbit

Quality Level

form

solid

shipped in

wet ice

storage temp.

−20°C

Application

Muscle acetone powder from rabbit has been used for the isolation of G-actin or actin.

Preparation Note

Prepared by a modification of Carsten, M.E. and Mommaerts, F.H., Biochemistry, 2, 28 (1963).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G-actin guides p53 nuclear transport: potential contribution of monomeric actin in altered localization of mutant p53
Saha T, et al.
Scientific reports, 6(4), 32626-32626 (2016)
Crosslinking activity of non-muscle myosin II is not sufficient for embryonic cytokinesis in C. elegans
Osorio D S, et al.
Development, 146(21), 32626-32626 (2019)
Taniya Saha et al.
Scientific reports, 6, 32626-32626 (2016-09-08)
p53 preserves genomic integrity by restricting anomaly at the gene level. Till date, limited information is available for cytosol to nuclear shuttling of p53; except microtubule-based trafficking route, which utilizes minus-end directed motor dynein. The present study suggests that monomeric
Interactome and F-actin interaction analysis of Dictyostelium discoideum coronin A
Fabrice T, et al.
International Journal of Molecular Sciences, 21(4), 1469-1469 (2020)
Daniel S Osório et al.
Development (Cambridge, England), 146(21) (2019-10-05)
Cytokinesis in animal cells requires the assembly and constriction of a contractile actomyosin ring. Non-muscle myosin II is essential for cytokinesis, but the role of its motor activity remains unclear. Here, we examine cytokinesis in C. elegans embryos expressing non-muscle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service