Skip to Content
Merck
All Photos(1)

Key Documents

M046000

Sigma-Aldrich

Cyanine 5 Phosphoramidite

Synonym(s):

2-[5-[1,3-Dihydro-1-[3-(4-methoxytriphenylmethoxy)propyl]-3,3-dimethyl-2H-indol-2-ylidene]-1,3-pentadien-1-yl]-1-[3-[N,N-diisopropylamino(2-cyanoethoxy)phosphinoxy]propyl]-3,3-dimethyl-3H-indolium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H72N4O4P
CAS Number:
Molecular Weight:
944.21
UNSPSC Code:
41116105

product line

Proligo Reagents

Assay

≥90% (31P-NMR)
≥95.0% (reversed phase HPLC)

drug control

USDEA Schedule I

color

clear blue0.1M in Acetonitrile

shipped in

dry ice
dry ice

storage temp.

−20°C

SMILES string

CC(C1=C(N/2CCCOP(N(C(C)C)C(C)C)OCCC#N)C=CC=C1)(C)C2=C\C=C\C=C\C(C3(C)C)=[N+](CCCOC(C4=CC=CC=C4)(C5=CC=C(OC)C=C5)C6=CC=CC=C6)C7=C3C=CC=C7.[Cl-]

InChI

1S/C60H72N4O4P.ClH/c1-46(2)64(47(3)4)69(67-44-23-40-61)68-45-25-42-63-55-33-22-20-31-53(55)59(7,8)57(63)35-18-12-17-34-56-58(5,6)52-30-19-21-32-54(52)62(56)41-24-43-66-60(48-26-13-10-14-27-48,49-28-15-11-16-29-49)50-36-38-51(65-9)39-37-50;/h10-22,26-39,46-47H,23-25,41-45H2,1-9H3;1H/q+1;/p-1

InChI key

GWCNEDQILVVBDJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Phosphoramidite technology is ideally suited for the modification of synthetic oligonucleotides with covalently attached reporter moieties, linkers, spacers, or haptens. Sigma-Aldrich provides popular reporters and linkers ready to use for DNA/RNA synthesis machines. Cyanine 5 Phosphoramidite is one such label whose absorbance maxima is 646 nm and emission maxima is 662 nm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service