Skip to Content
Merck
All Photos(1)

Key Documents

IAK0110

Sigma-Aldrich

Zymosan A Fluorescent Particles

Green Zymosan conjugate for phagocytosis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.47
Pricing and availability is not currently available.

Quality Level

concentration

1 mg/mL

suitability

suitable for fluorescence

storage temp.

2-8°C

General description

Phagocytosis is a cellular process for ingesting and eliminating microorganisms, foreign substances, and apoptotic cells. It is a specific form of endocytosis initiated by recognition and binding of foreign particles by cell surface receptors, followed by their engulfment, and formation of phagosomes. Maturing phagosomes transform to phagolysosomes which destroy the pathogen through enzymes and toxic peroxides. Zymosan prepared from yeast cell wall (Saccharomyces cerevisiae) and consisting of protein-carbohydrate complexes is frequently used as a pathogen in phagocytosis assays.

Application

Zymosan A fluorescent particles were prepared by conjugating Zymosan A particles with a bright green fluorescent molecular probe. The particles are ready-to-use phagocytosis-inducing particles for fluorescent imaging, microplate reader (Ex/Em = 490/520 nm) and flow cytometric analysis of phagocytosis. We recommend using 5 μL of Zymosan A fluorescent particles per 5 x 105 of viable cells.

Suitability

Suitable for inducing phagocytosis and screening activators and/or inhibitors of phagocytosis and Toll-like receptor ligands (TLR) in mammalian cells.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service