Merck
All Photos(4)
Key Documents
HS120018
Work 2 Store® Expanding Tube Storage Box
white, holds 64 x 1.5/2.0 mL Tubes
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
Pricing and availability is not currently available.
material
polycarbonate
polypropylene
white
feature
holds 64 x 1.5/2.0 mL Tubes
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120018
L × W × H
310 mm × 128 mm × 34 mm
General description
The Work 2 Store is an expandable single rack and storage box in one. Reduce sample handling time and the need for several racks when working with samples for cold temperature storage. The opening and closing mechanism allows the rack to expand, creating needed room for sample manipulation. When done, simply collapse the rack back to its compact form and it′s ready for the freezer. The Work 2 Store holds 1.5/2.0 mL cryo and microtubes.
Features and Benefits
  • Expanding wells have a center depression to stabilize conical tubes and a cryogenic vial lock for one handed cap removal
  • Holds 64 x 1.5/2.0 mL cryo and/or microtubes
  • Sides made from polycarbonate, Tube holders made of polypropylene
  • Instantly creates more working space
  • Helps reduce potential cross contamination
  • Stops opened microfuge lids from interfering with other wells
  • Stackable
  • Alphanumeric identification for quick location and retrieval of tubes
  • Clear lid locks the rack together for safe handling and transportation
  • Will fit into standard 2" (53mm) freezer racking
  • Temperature range -80 to 121ºC. Must use tubes capable of being stored at the required temperature of the user
  • Autoclavable
Legal Information
Work 2 Store is a registered trademark of Heathrow Scientific LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)